missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOPC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12163-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
PDZ domains contain approximately 90 amino acids and bind the extreme C terminus of proteins in a sequence-specific manner. PIST, a PDZ domain-containing Golgi protein, was discovered in a yeast two-hybrid system as a binding partner to Beclin-1, a Bcl-2-interacting protein homologous to the yeast autophagy gene apg6. Experiments with mutant PIST proteins lacking the PDZ domain showed that PIST interaction with Beclin-1 through its coiled-coil domain can modulate Beclin-1 activity and suggest that PIST interactions with other proteins through its PDZ domain may regulate the activity of PIST and Beclin-1.Specifica
GOPC | |
Polyclonal | |
Unconjugated | |
GOPC | |
CAL, CFTR associated ligand, FIG, Fused in glioblastoma, GOPC, GOPC1, PIST | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
309774, 57120, 94221 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BH60, Q9HD26 | |
GOPC | |
GOPC Fusion Protein Ag2804 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |