Learn More
gp100 / PMEL17 / SILV Rabbit, Clone: PMEL/1825R, enQuireBio™
Rabbit Monoclonal Antibody
Marca: EnquireBio 6490-RBM5-P1
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
gp100 / PMEL17 / SILV | |
Cytoplasmic | |
PMEL/1825R | |
Unconjugated | |
NP_001307051 | |
Rabbit | |
90-100kDa | |
Protein A/G Purified | |
RUO | |
6490 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
SILV | |
Recombinant full-length human SILV protein | |
SK-MEL-28 cells or Melanoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |