Learn More
GPR82 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA3039
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
IHC(P) analysis shows positive staining of GPR82 in human kidney tubules and germinal center of human lymph node. WB analysis shows GPR82 in glyco-protein enriched fractions from mock transfected or GPR82 overexpressed 293 cells. GPR82 exists in different glycosylated forms which may cause the molecular weight detected by WB to vary.
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.Specifica
GPR82 | |
Polyclonal | |
Unconjugated | |
GPR82 | |
G protein-coupled receptor 82; GPR82; Probable G-protein coupled receptor 82 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
Q96P67 | |
GPR82 | |
KLH conjugated COOH-terminal amino acids of 317-336 (KKTLYNLFTKSNSAHMQSYG) | |
RUO | |
27197 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |