Learn More
Thermo Scientific Chemicals Grade 25 Sapphire Spheres, Dia.: 4mm
Marca: Thermo Scientific™ 042059.KH
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Sapphire Sphere | |
4 mm | |
Grade 25 | |
2 Pieces |
RUO – Research Use Only