missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRO alpha Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12335-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The GRO gene was originally identified by subtractive hybridization studies between normal and tumorigenic Chinese hamster embryo fibroblasts. The hamster cDNA was cloned and used as a probe for cloning of the human GRO cDNA. The GROalpha gene initially cloned from T24 cells and the gene in melanoma cells encoding melanoma growth stimulating protein (MGSA) are identical.Specifica
GRO alpha | |
Polyclonal | |
Unconjugated | |
CXCL1 | |
C X C motif chemokine 1, CXCL1, FSP, GRO, GRO alpha(1 73), GRO1, GROa, Growth regulated alpha protein, MGSA, MGSA a, NAP 3, SCYB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2919 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09341 | |
CXCL1 | |
GRO alpha Fusion Protein Ag2917 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |