Learn More
GSDMB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA554929
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: AGGDMIAVRS LVDADRFRCF HLVGEKRTFF GCRHYTTGLT LMDILDTDGD KWLDELDSGL QGQKAEFQIL DNVDSTGELI VRLPKEIT Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 33%.
GSDMB Polyclonal antibody specifically detects GSDMB in Human samples. It is validated for Immunohistochemistry.Specifica
GSDMB | |
Polyclonal | |
Unconjugated | |
GSDMB | |
gasdermin B; gasdermin-B; Gasdermin-like protein; GSDMB; GSDML; PP4052; PRO2521 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55876 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TAX9 | |
GSDMB | |
Recombinant protein corresponding to Human GSDMB | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only