Learn More
Abnova™ GST Full-length ORF (AAB37352, 1 a.a. - 242 a.a.) Protein
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ P0002.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: MESPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKLTQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEMLKMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKSSKYIAWPLQGWQATFGGGDHPPKSDLEVLFQGPLEDPGYRGRTSFVSpecifica
AAB37352 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
GST Full-length ORF (AAB37352, 1 a.a. - 242 a.a.) Protein | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at −80°C. Aliquot to avoid repeated freezing and thawing. | |
GST | |
GST Tag |
Protein Array, ELISA, Western Blot | |
26.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10μg | |
RUO | |
Wheat Germ (in vitro) | |
wheat germ expression system |