missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTT2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17622-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Specifica
GSTT2B | |
Polyclonal | |
Unconjugated | |
GSTT2B | |
GST class theta 2, GSTT2, GSTT2B, GSTT2P | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
653689 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P0CG30 | |
GSTT2B | |
GSTT2B Fusion Protein Ag11828 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |