Learn More
H-B Instrument™ Durac™ Blood Bank Thermometers
Indicate critical temperature ranges in blood banks
Marca: H-B Instrument™ B60805-0100
Dettagli aggiuntivi : Peso : 0.02490kg Transport : UN number : 1090 Chem class : 3 Pack group : II
Descrizione
- Individually serialized
- Critical temperatures (4°C and 6°C) marked with arrows
- Recyclable packaging
Specifica
Total Immersion | |
152 mm | |
Red Organic Liquid | |
20°C | |
-5°C to +20°C | |
1°C |
6 in. | |
Glass | |
Blood Bank Thermometer, PFA coated | |
-5°C | |
±1°C |