Learn More
Goat anti-Syrian Hamster IgG (H+L) Cross-Adsorbed Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Marca: Invitrogen SA510284
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
This antibody will react with hamster IgG and with light chains common with other hamster immunoglobulins as determined by ELISA and IEP techniques. It has less than 1% cross reactivity with mouse and rat IgG. This antibody may cross react with IgG from other species.
Syrian Hamster IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Syrian Hamster IgG (H+L) Cross-Adsorbed in Hamster samples. It is validated for ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot.Specifica
Syrian Hamster IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Hamster | |
Liquid |
ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Hamster IgG h+l | |
1 mg | |
Secondary | |
4° C | |
IgG |