Learn More
IKA™ HELISTAND Electrical Viscometer Stand
The HELISTAND Electrical Viscometer Stand measures the viscosity of highly viscous samples with a high level of reliability. Designed for use with IKA™ ROTAVISC Rotational Viscometers.
Marca: IKA™ 0025007567
Dettagli aggiuntivi : Peso : 7.03000kg
Descrizione
- Electrical stand for IKA™ ROTAVISC Rotational Viscometers
Specifica
Viscometer Stand | |
IKA™ ROTAVISC Viscometer | |
EU Plug |