Learn More
IKA™ HELISTAND Electrical Viscometer Stand Set
The HELISTAND Electrical Viscometer Stand Set measures the viscosity of highly viscous samples with a high level of reliability. Includes a set of T-spindles. Designed for use with IKA™ ROTAVISC Rotational Viscometers.
Marca:  IKA™ 0025008233
Dettagli aggiuntivi : Peso : 6.00000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17227569
  • 1630.00€ / Pezzo
Spedizione stimata: 07-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Electrical stand for IKA™ ROTAVISC Rotational Viscometers
  • Accommodates substances so viscous that would be forced to the edges of a sample container by a static viscometer through slow vertical movements
  • Generates meaningful viscosity measurements when combined with the included T-spindles for non-flowing samples
Specifica
Specifica
Viscometer Stand
Includes T-Spindle Set
EU Plug
IKA™ ROTAVISC Viscometer
Viscosity measurement of non-flowing liquids