Learn More
Cytiva (formerly Pall Lab) HEPA Capsule with Versapor™ Membrane
Capsule ensures high efficiency particulate-free air (HEPA).
Marca: Cytiva (formerly Pall Lab) 12144
Dettagli aggiuntivi : Peso : 0.20000kg
Descrizione
- Porous membrane delivers high air flow rates at low differential pressure and extends filter life
- Ensures particulate-free air 99.97% retention of 0.3 μm DOP aerosol
- Designed for laboratory applications only
Specifica
Acrylic Copolymer on a Non-woven Support | |
Sterile | |
3/8 in. FNPT and Hose Barb |
1.2 μm | |
860 cm2 |