missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HER-2 / c-erbB-2 / neu / CD340 Mouse, Clone: ERBB2/2452, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 2064-MSM12-P1
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
HER-2 / c-erbB-2 / neu / CD340 | |
Cell Surface | |
ERBB2/2452 | |
Unconjugated | |
Mouse | |
185kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
2064 | |
IgG2a κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human HER-2 protein fragment (apprx. aa 311-462) | |
SKBR-3 Cells or Breast Cancers. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |