Learn More
Syngene™ Hi Intensity LEDS for GBOX
Syngene™ Hi Intensity LEDS for GBOX includes gantry, electronics and lighting units, and works with latest GBOX ONLY (not previous modular system).
Marca: Syngene™ HILED-GBIRFR
Descrizione
- For use with GBOX
- Includes gantry, electronics and lighting units
Specifica
Multiplex Western Fluorescence | |
1 | |
Gbox Chemi Systems | |
Green, Blue, Infa Red, Far Red |