missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scotch-Brite™ High Performance Microfibre Wipes 2010
High performance microfibre cloth that can be used on all surfaces, even delicate ones.
Marca: Scotch-Brite™ 7000103436
Dettagli aggiuntivi : Peso : 0.60000kg
Descrizione
- High performance microfibre wipe offering highly efficient, and lint free cleaning on virtually any surface.
- High quality cleaning results even when used with or without water, and with or without detergent
- Extremely durable, and can resist up to 500 wash cycles
- Machine washable up to 95°C
Specifica
360 mm | |
White | |
5/Pk. |
320 mm | |
320 x 360 mm |