missing translation for 'onlineSavingsMsg'
Learn More

HIST1H1T Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18188-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898244

  • 121.38€ / 20µL

Spedizione stimata: 28-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element.
Specifica

Specifica

HIST1H1T
Polyclonal
Unconjugated
HIST1H1T
H1FT, H1t, HIST1H1T, histone cluster 1, H1t, Histone H1t, Testicular H1 histone
Rabbit
Antigen Affinity Chromatography
RUO
107970, 3010
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P22492, Q07133
Hist1h1t
HIST1H1T Fusion Protein Ag12908
20 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali