missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone Deacetylase 2/HDAC2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-41285
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Histone Deacetylase 2/HDAC2 Polyclonal specifically detects Histone Deacetylase 2/HDAC2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifica
| Histone Deacetylase 2/HDAC2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| HDAC2 | |
| Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human HDAC2. The immunogen is located within the last 50 amino acids of HDAC2. Amino Acid Squence: KKARIEEDKKETEDKKTD | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least two isoforms of HDAC2 are known to exist; this antibody will detect both isoforms. HDAC2 antibody is predicted to not cross-react with other members of the HDAC family. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5-1 ug/ml, ELISA, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence 20 ug/ml | |
| EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1 | |
| Rabbit | |
| 52 kDa | |
| 0.1 mg | |
| Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases, Stem Cell Markers | |
| 3066 | |
| Human, Mouse, Rat | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto