Learn More
Histone H1.2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-49202
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Histone H1.2 Polyclonal antibody specifically detects Histone H1.2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifica
| Histone H1.2 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| H1 histone family, member 2, H1.2, H1F2H1C, histone 1, H1c, histone cluster 1, H1c, histone H1.2, Histone H1d, MGC3992 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KKAGATAGKRKASGPPVSELITKAVAA | |
| 0.1 mL | |
| Epigenetics | |
| 3006 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |