missing translation for 'onlineSavingsMsg'
Learn More

HLA class I ABC Mouse anti-Human, Porcine, Clone: 5C5B7, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66013-1-IG-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897505

  • 121.38€ / 20µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

HLA-class I major histocompatibility (MHC) antigens are intrinsic membrane glycoproteins expressed on nucleated cells and noncovalently associated with an invariant beta2 microglobulin. They carry foreign determits important for immune recognition by cytotoxic T cells, thus important for anti-viral and anti-tumour defence. Human HLA-class I antigens are represented by HLA-A, HLA-B and HLA-C molecules.
Specifica

Specifica

HLA class I ABC
Monoclonal
1.11 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P30443
HLA-A, LOC100515902
HLA class I ABC Fusion Protein Ag7488
20 μL
Primary
Human, Porcine
Antibody
IgG2b
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
5C5B7
Unconjugated
HLA-A
HLA A, HLA class I, HLA class I (HLA-A), HLA class I ABC, HLA-A, MHC class I antigen A*3
Mouse
Protein A
RUO
100515902, 3105
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali