missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11874-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
In mammals, histamine is metabolized by two major pathways: N(tau)-methylation via histamine N-methyltransferase and oxidative deamination via diamine oxidase. HNMT is the first enzyme which is found in the cytosol and uses S-adenosyl-L-methionine as the methyl donor. In the mammalian brain, the neurotransmitter activity of histamine is controlled by N(tau)-methylation as diamine oxidase is not found in the central nervous system. A common genetic polymorphism affects the activity levels of this gene product in red blood cells.Specifica
HNMT | |
Polyclonal | |
Unconjugated | |
HNMT | |
histamine N methyltransferase, HMT, HNMT, HNMT S1, HNMT S2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140483, 3176, 81676 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P50135, Q01984, Q91VF2 | |
HNMT | |
HNMT Fusion Protein Ag2462 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |