Learn More
Fisherbrand™ Horizontal Large Gel Systems: Combs
For large gel systems
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.22000kg
Descrizione
For large gel systems
Specifica
Owl™ A2 and A5 Large Gel Electrophoresis System | |
25 x 20 cm | |
0.1 cm | |
9.8 in. | |
7.8 in. | |
20 cm |
Standard Comb | |
8 | |
110 μL | |
25 cm | |
0.88 in. | |
2.25 cm |