missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18210-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.Specifica
HOXA6 | |
Polyclonal | |
Unconjugated | |
HOXA6 | |
homeobox A6, Homeobox protein Hox 1B, Homeobox protein Hox A6, HOX1, HOX1.2, HOX1B, HOXA6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15403, 3203 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09092, P31267 | |
HOXA6 | |
HOXA6 Fusion Protein Ag12875 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |