missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HPCA (Human) Recombinant Protein
Human HPCA full-length ORF ( NP_002134, 1 a.a. - 193 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00003208-P01.25ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Sequence: MGKQNSKLRPEMLQDLRENTEFSELELQEWYKGFLKDCPTGILNVDEFKKIYANFFPYGDASKFAEHVFRTFDTNSDGTIDFREFIIALSVTSPGRLEQKLMWAFSMYDLDGNGYISREEMLEIVQAIYKMVSSVMKMPEDESTPEKRTEKIFRQMDTNNDGKLSLEEFIRGAKSDPSIVRLLQCDPSSRSQF
Specifica
NP_002134 | |
hippocalcin | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HPCA | |
GST |
3208 | |
Wheat germ expression system | |
25 μg | |
BDR2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |