Learn More
Cytiva HR 5 Filter
Marca: Cytiva 18-6353-01
Dettagli aggiuntivi : Peso : 1.00000kg
Specifica
Adapter for HR 5 columns | |
10 filters and 4 o-rings | |
Filter HR 5 | |
Replacing the filter in the PTFE ring on the adaptor for HR 5 columns |