missing translation for 'onlineSavingsMsg'
Learn More

HSPA1L Mouse anti-Human, Mouse, Rat, Clone: 3E7H2, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66780-1-IG-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16808975

  • 140.00€ / 20µL
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This antibody well recognizes the endogenous HSPA1L protein in multiple cell lines.

In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock proteins. One class of stress proteins, termed the "small heat shock proteins" is comprised of a diverse group of proteins from ∽15 to >30 kDa, with a single form found in yeast and human cells and multiple related forms found in higher plants. A direct relationship between elevated stress protein expression and neoplasia has been suggested from studies on estrogen-induced proteins in breast cancer cells. In addition, Hsp27 exhibits rapid increased phosphorylation in response to various mitogens, tumor promoter (e. g. phorbol esters) and calcium ionophores. The expression of a 24 kDa protein was shown to correlate with steroid hormone receptor in cell lines, organs, and tumors. The 24 kDa estrogen-induced protein was found to be expressed at high levels in carcinoma of the breast. Another study has suggested that levels of the Hsp27 correlate with endometrial adenocarcinomas. Interestingly, the regulation of human Hsp27 by estrogens and heat shock is reminiscent of the ecdysteroid and heat shock activation of the drosophila Hsp20 genes.
Specifica

Specifica

HSPA1L
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P16627, P34931, P55063
Hspa1l
HSPA1L Fusion Protein Ag5182
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG2b
Western Blot
3E7H2
Unconjugated
HSPA1L
Heat shock 70 kDa protein 1L, HSP70 1L, HSP70 HOM, HSP70T, HSPA1L, hum70t
Mouse
Protein A
RUO
15482, 24963, 3305
-20°C
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali