Learn More
Human Acetylcholinesterase/ACHE Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marca: R&D Systems AF7574-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Acetylcholinesterase/ACHE Polyclonal specifically detects Acetylcholinesterase/ACHE in Human samples. It is validated for Immunohistochemistry.
Specifica
| Acetylcholinesterase/ACHE | |
| Polyclonal | |
| Immunohistochemistry 3-15 ug/mL | |
| P22303 | |
| Sheep | |
| Antigen Affinity-purified | |
| RUO | |
| 43 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group | |
| Chinese hamster ovary cell line CHO-derived recombinant human ACHE Met1-Leu614 Accession # P22303 | |
| 25 μg | |
| Primary | |
| Detects human ACHE in direct ELISAs. | |
| Human | |
| IgG |