Learn More
Human Acetylcholinesterase/ACHE Antibody, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody
251.00€ - 603.00€
Specifica
Antigene Acetylcholinesterase/ACHE
Diluizione Immunohistochemistry 3-15 ug/mL
Applicazioni Immunohistochemistry
Classificazione Polyclonal
Coniugato Unconjugated
Visualizza altre specifiche
Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
18768417
GREENER_CHOICE
Vedere i documenti
R&D Systems
AF7574-SP
25 μg
251.00€
25µg
Visualizzare prodotti alternativi
Risparmia fino a 
»
18735367
GREENER_CHOICE
Vedere i documenti
R&D Systems
AF7574-100
100 μg
603.00€
100µg
Visualizzare prodotti alternativi
Risparmia fino a 
»
Descrizione
Descrizione
Acetylcholinesterase/ACHE Polyclonal specifically detects Acetylcholinesterase/ACHE in Human samples. It is validated for Immunohistochemistry.
Specifica
Specifica
Acetylcholinesterase/ACHE
Immunohistochemistry
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group
Chinese hamster ovary cell line CHO-derived recombinant human ACHE Met1-Leu614 Accession # P22303
Primary
Reconstitute at 0.2 mg/mL in sterile PBS.
Detects human ACHE in direct ELISAs.
Immunohistochemistry 3-15 ug/mL
Polyclonal
Sheep
Human
P22303
43
IgG
Antigen Affinity-purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.