Learn More
Abnova™ Human ADAM22 Full-length ORF (AAH36029.1, 1 a.a. - 342 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00053616-P01.25ug
Descrizione
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is highly expressed in the brain and may function as an integrin ligand in the brain. Alternative splicing results in several transcript variants. [provided by RefSeq]
Sequence: MQAAVAVSVPFLLLCVLGTCPPARCGQAGDASLMELEKRKENRFVERQSIVPLRLIYRSGGEDESRHDALDTRVRGDVGGPQLTHVDQASFQVDAFGTSFILDVVLNHDLLSSEYIERHIEHGGKTVEVKGGEHCYYQGHIRGNPDSFVALSTCHGLHGMFYDGNHTYLIEPEENDTTQEDFHFHSVYKSRLFEFSLDDLPSEFQQVNITPSKFILKPRPKRSKRQLRRYPRNVEEETKYIELMIVNDHLMFKKHRLSVVHTNTYAKSVVNMADLIYKDQLKTRIVLVAMETWATDNKFAISENPLITLREFMKYRRDFIKEKSDAVHLFSYVTSVMMYYFFSpecifica
AAH36029.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 μg | |
RUO | |
ADAM22 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
53616 | |
ADAM22 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MDC2/MGC149832 | |
ADAM22 | |
Recombinant | |
wheat germ expression system |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.