missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ADAMTS15 Partial ORF (NP_620686, 863 a.a. - 950 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00170689-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16132327

  • 508.00€ / 25µg
Spedizione stimata: 26-08-2024
per visualizzare lo stock disponibile

Descrizione

Descrizione

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has a high sequence similarity to the proteins encoded by ADAMTS1 and ADAMTS8. [provided by RefSeq]

Sequence: AVDCRGSAGQRTVPACDAAHRPVETQACGEPCPTWELSAWSPCSKSCGRGFQRRSLKCVGHGGRLLARDQCNLHRKPQELDFCVLRPC
Specifica

Specifica

NP_620686
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.42kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AVDCRGSAGQRTVPACDAAHRPVETQACGEPCPTWELSAWSPCSKSCGRGFQRRSLKCVGHGGRLLARDQCNLHRKPQELDFCVLRPC
RUO
ADAMTS15
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
170689
ADAMTS15 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC126403
ADAMTS15
Recombinant
wheat germ expression system
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali