missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AK5 Partial ORF (NP_777283, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00026289-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16125466

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a member of the adenylate kinase family, which is involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate groups among adenine nucleotides. This member is related to the UMP/CMP kinase of several species. It is located in the cytosol and expressed exclusively in brain. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MNTNDAKEYLARREIPQLFESLLNGLMCSKPEDPVEYLESCLQKVKELGGCDKVKWDTFVSQEKKTLPPLNGGQSRRSFLRNVMPENSNFPYRRYDRLPP
Specifica

Specifica

NP_777283
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MNTNDAKEYLARREIPQLFESLLNGLMCSKPEDPVEYLESCLQKVKELGGCDKVKWDTFVSQEKKTLPPLNGGQSRRSFLRNVMPENSNFPYRRYDRLPP
RUO
AK5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
26289
AK5 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AK6/MGC33326
AK5
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali