missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ALKBH Full-length ORF (AAH25787.1, 1 a.a. - 389 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008846-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129445

  • 349.00€ / 10µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a homolog to the E. coli alkB gene product. The E. coli alkB protein is part of the adaptive response mechanism of DNA alkylation damage repair. It is involved in damage reversal by oxidative demethylation of 1-methyladenine and 3-methylcytosine. [provided by RefSeq]

Sequence: MGKMAAAVGSVATLATEPGEDAFRKLFRFYRQSRPGTADLEGVIDFSAAHAARGKGPGAQKVIKSQLNVSSVSEQNAYRAGLQPVSKWQAYGLKGYPGFIFIPNPFLPGYQWHWVKQCLKLYSQKPNVCNLDKHMSKEETQDLWEQSKEFLRYKEATKRRPRSLLEKLRWVTVGYHYNWDSKKYSADHYTPFPSDLGFLSEQVAAACGFEDFRAEAGILNYYRLDSTLGIHVDRSELDHSKPLLSFSFGQSAIFLLGGLQRDEAPTAMFMHSGDIMIMSGFSRLLNHAVPRVLPNPEGEGLPHCLEAPLPAVLPRDSMVEPCSMEDWQVCASYLKTARVNMTVRQVLATDQNFPLEPIEDEKRDISTEGFCHLDDQNSEVKRARINPHS
Specifica

Specifica

AAH25787.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
68.42kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ALKBH1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
8846
ALKBH (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ABH/ABH1/ALKBH/alkB/hABH
ALKBH1
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali