missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ALS2CR8 Full-length ORF (NP_079020.12, 1 a.a. - 718 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00079800-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16124493

  • 528.00€ / 25µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Sequence: MEQSNDSLRVNHNDGEESKTSAQVFEHLICMDSRDSSFGQNDSPTVLPITTREANNSLISQNIPGPLTQTQTLSAEQFHLVDQNGQAIQYELQSLGESNAQMMIVASPTENGQVLRVIPPTQTGMAQVIIPQGQLVDVNSPRDVPEEKPSNRNLPTVRVDTLADNTSNYILHPQTSFPLPKKSVTGMLEEPLLGPLQPLSSNTPIWACRLRSCEKIGDSYRGYCVSETELESVLTFHKQQTQSVWGTRQSPSPAKPATRLMWKSQYVPYDGIPFVNAGSRAVVMECQYGPRRKGFQLKKVSEQESRSCQLYKATCPARIYIKKVQKFPEYRVPTDPKIDKKIIRMEQEKAFNMLKKNLVDAGGVLRWYVQLPTQQAHQYHELETPCLTLSPSPFPVSSLEEEETAVRDENCALPSRLHPQVAHKIQELVSQGIEQVYAVRKQLRKFVERELFKPDEVPERHNLSFFPTVNDIKNHIHEVQKSLRNGDTVYNSEIIPATLQWTTDSGNILKETMTVTFAEGNSPGESITTKVETNQTRGSLSPEPTHLLSSLSSFQPKIFTQLQGLQLQPRYTSPDESPAVVSVNNQPSSSPSGLLDTIGSAVMNNNSLLLGQSHSLQRDTCLTQNNSTASTMGNLPEPDQNLVAMDELVEVGDVEDTGNLEGTVHRILLGDVQTIPIQIIDNHSALIEENPESTISVSQVKQEPKEPALSMEAKNCGL
Specifica

Specifica

NP_079020.12
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
106.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ALS2CR8
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
79800
ALS2CR8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CARF/DKFZp667N246/FLJ12675/FLJ21579/NYD-SP24
ALS2CR8
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali