missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AMPD2 Full-length ORF (NP_631895.1, 1 a.a. - 798 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000271-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16171541

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Adenosine monophosphate deaminase-2 (EC 3.5.4.6) catalyzes the deamination of AMP to IMP and plays an important role in the purine nucleotide cycle.[supplied by OMIM]

Sequence: MASEARGGLGAPPLQSARSLPGPAPCLKHFPLDLRTSMDGKCKEIAEELFTRSLAESELRSAPYEFPEESPIEQLEERRQRLERQISQDVKLEPDILLRAKQDFLKTDSDSDLQLYKEQGEGQGDRSLRERDVLEREFQRVTISGEEKCGVPFTDLLDAAKSVVRALFIREKYMALSLQSFCPTTRRYLQQLAEKPLETRTYEQGPDTPVSADAPVHPPALEQHPYEHCEPSTMPGDLGLGLRMVRGVVHVYTRREPDEHCSEVELPYPDLQEFVADVNVLMALIINGPIKSFCYRRLQYLSSKFQMHVLLNEMKELAAQKKVPHRDFYNIRKVDTHIHASSCMNQKHLLRFIKRAMKRHLEEIVHVEQGREQTLREVFESMNLTAYDLSVDTLDVHADRNTFHRFDKFNAKYNPIGESVLREIFIKTDNRVSGKYFAHIIKEVMSDLEESKYQNAELRLSIYGRSRDEWDKLARWAVMHRVHSPNVRWLVQVPRLFDVYRTKGQLANFQEMLENIFLPLFEATVHPASHPELHLFLEHVDGFDSVDDESKPENHVFNLESPLPEAWVEEDNPPYAYYLYYTFANMAMLNHLRRQRGFHTFVLRPHCGEAGPIHHLVSAFMLAENISHGLLLRKAPVLQYLYYLAQIGIAMSPLSNNSLFLSYHRNPLPEYLSRGLMVSLSTDDPLQFHFTKEPLMEEYSIATQVWKLSSCDMCELARNSVLMSGFSHKVKSHWLGPNYTKEGPEGNDIRRTNVPDIRVGYRYETLCQELALITQAVQSEMLETIPEEAGITMSPGPQ
Specifica

Specifica

NP_631895.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
118.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
AMPD2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
271
AMPD2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AMPD2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali