missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human APITD1 Full-length ORF (NP_954988.1, 1 a.a. - 138 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00378708-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16130434

  • 528.00€ / 25µg
Spedizione stimata: 30-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene was identified in the neuroblastoma tumour suppressor candidate region on chromosome 1p36. It contains a TFIID-31 domain, similar to that found in TATA box-binding protein-associated factor, TAF(II)31, which is required for p53-mediated transcription activation. This gene was expressed at very low levels in neuroblastoma tumours, and was shown to reduce cell growth in neuroblastoma cells, suggesting that it may have a role in a cell death pathway. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq]

Sequence: MEEEAETEEQQRFSYQQRLKAAVHYTVGCLCEEVALDKEMQFSKQTIAAISELTFRQCENFAKDLEMFARHAKRTTINTEDVKLLARRSNSLLKYITDKSEEIAQINLERKAQKKKKSEDGSKNSRQPAEAGVVESEN
Specifica

Specifica

NP_954988.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
42.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
APITD1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
378708
APITD1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CENP-S/CENPS/MGC32686
APITD1
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali