missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human APOA5 Full-length ORF (AAI01790.1, 1 a.a. - 366 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00116519-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16127353

  • 349.00€ / 10µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene is an apolipoprotein and an important determinant of plasma triglyceride levels, a major risk factor for coronary artery disease. It is a component of high density lipoprotein and is highly similar to a rat protein that is upregulated in response to liver injury. This gene uses alternate polyadenylation sites and is located proximal to the apolipoprotein gene cluster on chromosome 11q23. [provided by RefSeq]

Sequence: MASMAAVLTWALALLSAFSATQARKGFWDYFSQTSGDKGRVEQIHQQKMAREPATLKDSLEQDLNNMNKFLEKLRPLSGSEAPRLPQDPVGMRRQLQEELEEVKARLQPYMAEAHELVGWNLEGLRQQLKPYTMDLMEQVALRVQELQEQLRVVGEDTKAQLLGGVDEAWALLQGLQSRVVHHTGRFKELFHPYAESLVSGIGRHVQELHRSVAPHAPASPARLSRCVQVLSRKLTLKAKALHARIQQNLDQLREELSRAFAGTGTEEGAGPDPQMLSEEVRQRLQAFRQDTYLQIAAFTRAIDQETEEVQQQLAPPPPGHSAFAPEFQQTDSGKVLSKLQARLDDLWEDITHSLHDQGHSHLGDP
Specifica

Specifica

AAI01790.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
67.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
APOA5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
116519
APOA5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APOA-V/APOAV/MGC126836/MGC126838/RAP3
APOA5
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali