missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AREG Partial ORF (AAH09799, 20 a.a. - 129 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000374-Q01.25ug

 vedi altre versioni di questo prodotto

Codice prodotto. 16124824

  • 508.00€ / 25µg

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso

Descrizione

Descrizione

The protein encoded by this gene is a member of the epidermal growth factor family. It is an autocrine growth factor as well as a mitogen for astrocytes, Schwann cells, and fibroblasts. It is related to epidermal growth factor (EGF) and transforming growth factor alpha (TGF-alpha). This protein interacts with the EGF/TGF-alpha receptor to promote the growth of normal epithelial cells and inhibits the growth of certain aggressive carcinoma cell lines. This encoded protein is associated with a psoriasis-like skin phenotype. [provided by RefSeq]

Sequence: SGHYAAGLDLNDTYSGKREPFSGDHSADGFEVTSRSEMSSGSEISPVSEMPSSSEPSSGADYDYSEEYDNEPQIPGYIVDDSVRVEQVVKPPQNKTESENTSDKPKRKKK
Specifica

Specifica

AAH09799
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SGHYAAGLDLNDTYSGKREPFSGDHSADGFEVTSRSEMSSGSEISPVSEMPSSSEPSSGADYDYSEEYDNEPQIPGYIVDDSVRVEQVVKPPQNKTESENTSDKPKRKKK
RUO
AREG
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
374
AREG (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AR/CRDGF/MGC13647/SDGF
AREG
Recombinant
wheat germ expression system
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali