missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARHGAP4 Partial ORF (AAH52303, 881 a.a. - 986 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000393-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16171841

  • 349.00€ / 10µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a member of the rhoGAP family of proteins which play a role in the regulation of small GTP-binding proteins belonging to the RAS superfamily. The protein encoded by the orthologous gene in rat is localized to the Golgi complex and can redistribute to microtubules. The rat protein stimulates the activity of some Rho GTPases in vitro. Genomic deletions of this gene and a neighboring gene have been found in patients with nephrogenic diabetes insipidus. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: TSPEAMGPSGHRRRCLVPASPEQHVEVDKAVAQNMDSVFKELLGKTSVRQGLGPASTTSPSPGPRSPKAPPSSRLGRNKGFSRGPGAPASPSASHPQGLDTTPKPH
Specifica

Specifica

AAH52303
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.29kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TSPEAMGPSGHRRRCLVPASPEQHVEVDKAVAQNMDSVFKELLGKTSVRQGLGPASTTSPSPGPRSPKAPPSSRLGRNKGFSRGPGAPASPSASHPQGLDTTPKPH
RUO
ARHGAP4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
393
ARHGAP4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C1/KIAA0131/RGC1/RhoGAP4/p115
ARHGAP4
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali