missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARSF Full-length ORF (NP_004033.2, 1 a.a. - 590 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000416-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16131921

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene is a member of the sulfatase family, and more specifically, the arylsulfatase subfamily. Members of the subfamily share similarity in sequence and splice sites, and are clustered together on chromosome X, suggesting that they are derived from recent gene duplication events. Sulfatases are essential for the correct composition of bone and cartilage matrix. The activity of this protein, unlike that of arylsulfatase E, is not inhibited by warfarin. [provided by RefSeq]

Sequence: MRPRRPLVFMSLVCALLNTCQAHRVHDDKPNIVLIMVDDLGIGDLGCYGNDTMRTPHIDRLAREGVRLTQHISAASLCSPSRSAFLTGRYPIRSGMVSSGNRRVIQNLAVPAGLPLNETTLAALLKKQGYSTGLIGKWHQGLNCDSRSDQCHHPYNYGFDYYYGMPFTLVDSCWPDPSRNTELAFESQLWLCVQLVAIAILTLTFGKLSGWVSVPWLLIFSMILFIFLLGYAWFSSHTSPLYWDCLLMRGHEITEQPMKAERAGSIMVKEAISFLERHSKETFLLFFSFLHVHTPLPTTDDFTGTSKHGLYGDNVEEMDSMVGKILDAIDDFGLRNNTLVYFTSDHGGHLEARRGHAQLGGWNGIYKGGKGMGGWEGGIRVPGIVRWPGKVPAGRLIKEPTSLMDILPTVASVSGGSLPQDRVIDGRDLMPLLQGNVRHSEHEFLFHYCGSYLHAVRWIPKDDSGSVWKAHYVTPVFQPPASGGCYVTSLCRCFGEQVTYHNPPLLFDLSRDPSESTPLTPATEPLHDFVIKKVANALKEHQETIVPVTYQLSELNQGRTWLKPCCGVFPFCLCDKEEEVSQPRGPNEKR
Specifica

Specifica

NP_004033.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
92.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ARSF
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
416
ARSF (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASF
ARSF
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali