Learn More
ACROBiosystems Human ASGR1 / ASGPR1 Protein, Fc Tag
MALS verified
Marca: ACROBiosystems AS1-H5253-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
ASGPR, a transmembrane C-type lectin, recognizes a wide variety of ligands that contain either terminal galactose (Gal) or N-acetylgalactosamine (GalNAc) residues and has been identified as a highly selective receptor on hepatocytes. The ASGR is composed of both a major (ASGR1) and minor subunit (ASGR2). ASGR1 has been shown to be efficiently targeted to the plasma membrane and to undergo constitutive endocytosis and recycling and found mainly expressed in the human liver.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 52.8 kDa. The protein migrates as 65-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
NP_001662.1 | |
1 mg | |
Gln 62 - Leu 291 | |
ASGP-R 1, ASGPR 1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
52.8 kDa | |
Human ASGR1, Fc Tag (AS1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 62 - Leu 291 (Accession # P07306-1). | |
-20°C | |
ASGR1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |