Learn More
Abnova™ Human ASXL1 Full-length ORF (AAH64984.1, 1 a.a. - 84 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00171023-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene is similar to the Drosophila additional sex combs gene, which encodes a chromatin-binding protein required for normal determination of segment identity in the developing embryo. The protein is a member of the Polycomb group of proteins, which are necessary for the maintenance of stable repression of homeotic and other loci. The protein is thought to disrupt chromatin in localized areas, enhancing transcription of certain genes while repressing the transcription of other genes. The protein encoded by this gene functions as a ligand-dependent co-activator for retinoic acid receptor in cooperation with nuclear receptor coactivator 1. Mutations in this gene are associated with myelodysplastic syndromes and chronic myelomonocytic leukemia. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MKDKQKKKKERTWAEAARLVLENYSDAPMTPKQILQVIEAEGLKEMSGTSPLACLNAMLHSNSRGGEGLFYKLPGRISLFTLKRSpecifica
AAH64984.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ASXL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
171023 | |
ASXL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0978/MGC117280/MGC71111 | |
ASXL1 | |
Recombinant | |
wheat germ expression system |