Learn More
Abnova™ Human BANP Full-length ORF (NP_060339.2, 1 a.a. - 469 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00054971-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a protein that binds to matrix attachment regions. The protein functions as a tumor suppressor and cell cycle regulator. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: MMSEHDLADVVQIAVEDLSPDHPVVLENHVVTDEDEPALKRQRLEINCQDPSIKSFLYSINQTICLRLDSIEAKLQALEATCKSLEEKLDLVTNKQHSPIQVPMVAGSPLGATQTCNKVRCAVPGRRQNTIVVKVPGQEDSHHEDGESGSEASDSVSSCGQAGSQSIGSNVTLITLNSEEDYPNGTWLGDENNPEMRVRCAIIPSDMLHISTNCRTAEKMALTLLDYLFHREVQAVSNLSGQGKHGKKQLDPLTIYGIRCHLFYKFGITESDWYRIKQSIDSKCRTAWRRKQRGQSLAVKSFSRRTPNSSSYCPSEPMMSTPPPASELPQPQPQPQALHYALANAQQVQIHQIGEDGQVQVIPQGHLHIAQVPQGEQVQITQDSEGNLQIHHVGQDGQVLQGAQLIAVASSDPAAAGVDGSPLQGSDIQVQYVQLAPVSDHTAGAQTAEALQPTLQPEMQLEHGAIQIQSpecifica
NP_060339.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
BANP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
54971 | |
BANP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BEND1/DKFZp761H172/FLJ10177/FLJ20538/SMAR1/SMARBP1 | |
BANP | |
Recombinant | |
wheat germ expression system |