missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BTN1A1 Full-length ORF (AAH96312.1, 1 a.a. - 526 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000696-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16132381

  • 528.00€ / 25µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Butyrophilin is the major protein associated with fat droplets in the milk. It is a member of the immunoglobulin superfamily. It may have a cell surface receptor function. The human butyrophilin gene is localized in the major histocompatibility complex (MHC) class I region of 6p and may have arisen relatively recently in evolution by the shuffling of exons between 2 ancestral gene families [provided by RefSeq]

Sequence: MAVFPSSGLPRCLLTLILLQLPKLDSAPFDVIGPPEPILAVVGEDAELPCRLSPNASAEHLELRWFRKKVSPAVLVHRDGREQEAEQMPEYRGRATLVQDGIAKGRVALRIRGVRVSDDGEYTCFFREDGSYEEALVHLKVAALGSDPHISMQVQENGEICLECTSVGWYPEPQVQWRTSKGEKFPSTSESRNPDEEGLFTVAASVIIRDTSAKNVSCYIQNLLLGQEKKVEISIPASSLPRLTPWIVAVAVILMVLGLLTIGSIFFTWRLYNERPRERRNEFSSKERLLEELKWKKATLHAVDVTLDPDTAHPHLFLYEDSKSVRLEDSRQKLPEKTERFDSWPCVLGRETFTSGRHYWEVEVGDRTDWAIGVCRENVMKKGFDPMTPENGFWAVELYGNGYWALTPLRTPLPLAGPPRRVGIFLDYESGDISFYNMNDGSDIYTFSNVTFSGPLRPFFCLWSSGKKPLTICPIADGPERVTVIANAQDLSKEIPLSPMGEDSAPRDADTLHSKLIPTQPSQGAP
Specifica

Specifica

AAH96312.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
85.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
BTN1A1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
696
BTN1A1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BT/BTN
BTN1A1
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali