missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C15orf16 Partial ORF (NP_570971, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00161725-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16172247

  • 528.00€ / 25µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Deubiquitinating enzymes (DUBs; see MIM 603478) are proteases that specifically cleave ubiquitin (MIM 191339) linkages, negating the action of ubiquitin ligases. OTUD7A belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.[supplied by OMIM]

Sequence: MVSSVLPNPTSAECWAALLHDPMTLDMDAVLSDFVRSTGAEPGLARDLLEGKNWDLTAALSDYEQLRQVHTANLPHVFNEGRGPKQPEREPQPGHKVER
Specifica

Specifica

NP_570971
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MVSSVLPNPTSAECWAALLHDPMTLDMDAVLSDFVRSTGAEPGLARDLLEGKNWDLTAALSDYEQLRQVHTANLPHVFNEGRGPKQPEREPQPGHKVER
RUO
OTUD7A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
161725
C15orf16 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C15orf16/C16ORF15/CEZANNE2/OTUD7
OTUD7A
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali