Learn More
Abnova™ Human CACNG7 Partial ORF (NP_114102.2, 203 a.a. - 274 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00059284-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The mouse protein stargazin is one of five subunits comprising neuronal voltage-gated calcium channels. This subunit, gamma, is thought to stabilize the calcium channel in an inactive (closed) state. Mutations in the gene encoding stargazin have been associated with absence seizures, also known as petit-mal or spike-wave seizures. The protein encoded by this gene is structurally similar to the mouse stargazin protein and is a member of the neuronal calcium channel gamma subunit protein family. However, it appears unlikely that the encoded protein is part of a functional calcium channel. Rather, it appears to inhibit the expression of a specific calcium channel subunit. [provided by RefSeq]
Sequence: RYAEEEMYRPHPAFYRPRLSDCSDYSGQFLQPEAWRRGRSPSDISSDVSIQMTQNYPPAIKYPDHLHISTSPSpecifica
NP_114102.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.66kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RYAEEEMYRPHPAFYRPRLSDCSDYSGQFLQPEAWRRGRSPSDISSDVSIQMTQNYPPAIKYPDHLHISTSP | |
RUO | |
CACNG7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
59284 | |
CACNG7 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CACNG7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |