missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CAPN11 Partial ORF (NP_008989, 557 a.a. - 651 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00011131-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16126222

  • 349.00€ / 10µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Calpains constitute a family of intracellular calcium-dependent cysteine proteases. There are eight members in this superfamily. They consist of a variable 80 kDa subunit and an invariant 30 kDa subunit. This calpain protein appears to have protease activity and calcium-binding ability. A similar mouse protein may play a functional role in spermatogenesis and in the regulation of calcium-dependent signal transduction events during meiosis. [provided by RefSeq]

Sequence: LLNRMAIKFKSFKTKGFGLDACRCMINLMDKDGSGKLGLLEFKILWKKLKKWMDIFRECDQDHSGTLNSYEMRLVIEKAGIKLNNKVMQVLVARY
Specifica

Specifica

NP_008989
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.19kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LLNRMAIKFKSFKTKGFGLDACRCMINLMDKDGSGKLGLLEFKILWKKLKKWMDIFRECDQDHSGTLNSYEMRLVIEKAGIKLNNKVMQVLVARY
RUO
CAPN11
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
11131
CAPN11 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
calpain11
CAPN11
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali