missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CAPZB Partial ORF (NP_004921, 192 a.a. - 272 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000832-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16125644

  • 349.00€ / 10µg
Spedizione stimata: 30-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

CAPZB is a member of the F-actin capping protein family. This gene encodes the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments. [provided by RefSeq]

Sequence: SLTRQMEKDETVSDCSPHIANIGRLVEDMENKIRSTLNEIYFGKTKDIVNGLRSVQTFADKSKQEALKNDLVEALKRKQQC
Specifica

Specifica

NP_004921
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.65kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SLTRQMEKDETVSDCSPHIANIGRLVEDMENKIRSTLNEIYFGKTKDIVNGLRSVQTFADKSKQEALKNDLVEALKRKQQC
RUO
CAPZB
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
832
CAPZB (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAPB/CAPPB/CAPZ/MGC104401/MGC129749/MGC129750
CAPZB
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali