Learn More
ACROBiosystems Human CD161 Protein, Fc Tag
Marca:  ACROBiosystems CD1-H5253-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 17236080
  • dietro preventivo / 100µg
Spedizione stimata: 17-05-2024
per visualizzare lo stock disponibile
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
CD161 is also known as NKR-P1A, CLEC5B and KLRB1which plays an inhibitory role on natural killer (NK) cells cytotoxicity. Activation results in specific acid sphingomyelinase/SMPD1 stimulation with subsequent marked elevation of intracellular ceramide. Activation also leads to AKT1/PKB and RPS6KA1/RSK1 kinases stimulation as well as markedly enhanced T-cell proliferation induced by anti-CD3. Acts as a lectin that binds to the terminal carbohydrate Gal-alpha Gal epitope as well as to the N-acetyllactosamine epitope. Binds also to CLEC2D/LLT1 as a ligand and inhibits NK cell-mediated cytotoxicity as well as interferon-gamma secretion in target cells. New research shows that genetic inactivation of KLRB1 or antibody-mediated CD161 blockade enhances T cell-mediated killing of glioma cells in vitro and their anti-tumor function in vivo. KLRB1 and its associated transcriptional program are also expressed by substantial T cell populations in other human cancers. The reference provides an atlas of T cells in gliomas and highlights CD161 and other NK cell receptors as immunotherapy targets.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 44.9 kDa.
Specifica
Specifica
44.9 kDa
Human CD161, Fc Tag (CD1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 67 - Ser 225 (Accession # Q12918-1).
-20°C
CD161
Human
HEK293
Immune Checkpoint Proteins
100 μg
Gln 67 - Ser 225
CD161, C-type lectin domain family 5 member B, HNKR-P1a, NKR-P1A, Natural killer cell surface protein P1A
Unconjugated
Fc Tag
Powder