missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD20 (Research Grade Tositumomab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
Marca: R&D Systems MAB10440-100
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifica
| CD20 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 931 | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| Hu11A | |
| Flow Cytometry 0.25 ug/106 cells, CyTOF-ready | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35, Bp35MGC3969, CD20, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Ly-44, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A1, MS4A2, S7 | |
| Human CD20 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto