Learn More
Human CD20 (Research Grade Tositumomab Biosimilar) Antibody, R&D Systems™
Human Monoclonal Antibody
291.00€ - 644.00€
Specifica
| Antigene | CD20 |
|---|---|
| Clone | Hu11A |
| Diluizione | Flow Cytometry 0.25 ug/106 cells, CyTOF-ready |
| Applicazioni | Flow Cytometry, CyTOF |
| Classificazione | Monoclonal |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
18733537
![]() |
R&D Systems
MAB10440-SP |
25 μg |
291.00€
25µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
18723337
![]() |
R&D Systems
MAB10440-100 |
100 μg |
644.00€
100µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifica
| CD20 | |
| Flow Cytometry 0.25 ug/106 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 931 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| Hu11A | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Human | |
| Human | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35, Bp35MGC3969, CD20, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Ly-44, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A1, MS4A2, S7 | |
| Human CD20 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |